ENA061488412 Enamine

2-{[(5-methyl-1,2-oxazol-3-yl)methyl]sulfanyl}benzoic acid

  • MDL number MFCD06655907
  • Cas Number : 854357-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-161
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13526

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13526
Availability details for
Quantity
  • Availability
  • Add to Cart