ENA061488417 Enamine

methyl 2-(chloromethyl)-5-(2-methoxy-2-oxoethyl)-4-oxo-3,4-dihydrothieno[2,3-d]pyrimidine-6-carboxylate

  • MDL number MFCD06655909
  • Cas Number : 854357-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 208...210
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13528

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13528
Availability details for
Quantity
  • Availability
  • Add to Cart