ENA061488429 Enamine

3-bromo-2,3-dihydro-1lambda6-thiophene-1,1-dione

  • MDL number MFCD00462449
  • Cas Number : 53336-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54-55
Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13818

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13818
Availability details for
Quantity
  • Availability
  • Add to Cart