ENA061488430 Enamine

[(1,1-dioxido-1,2-benzisothiazol-3-yl)(methyl)amino]acetic acid

  • MDL number MFCD06655913
  • Cas Number : 854357-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13535

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13535
Availability details for
Quantity
  • Availability
  • Add to Cart