ENA061488431 Enamine

2-[2-(2H-1,2,3,4-tetrazol-5-yl)ethyl]-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD05260559
  • Cas Number : 412314-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13536

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13536
Availability details for
Quantity
  • Availability
  • Add to Cart