ENA061488440 Enamine

3-(2-oxo-2-phenylethyl)-5,6-dihydropyrazin-2(1H)-one

  • MDL number MFCD00169076
  • Cas Number : 1821-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13541

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13541
Availability details for
Quantity
  • Availability
  • Add to Cart