ENA061488442 Enamine

4-fluoro-3-(methoxymethyl)-1-benzothiophene-2-carboxylic acid

  • MDL number MFCD06655917
  • Cas Number : 854357-47-4
  • Molecular Weight : 240.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-139
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13542

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13542
Availability details for
Quantity
  • Availability
  • Add to Cart