ENA061488443 Enamine

5-[(diethylamino)methyl]quinolin-8-ol

  • MDL number MFCD06285492
  • Cas Number : 22506-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60-61
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13543

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13543
Availability details for
Quantity
  • Availability
  • Add to Cart