ENA061488446 Enamine

5-(4-fluorophenyl)-2-sulfanyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD06655918
  • Cas Number : 304684-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200-201
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-13546

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13546
Availability details for
Quantity
  • Availability
  • Add to Cart