ENA061488451 Enamine

N-(2-furylmethyl)-2-hydrazino-2-oxoacetamide

  • MDL number MFCD02307759
  • Cas Number : 91847-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157...159
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13840

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13840
Availability details for
Quantity
  • Availability
  • Add to Cart