ENA061488455 Enamine

2-[(2-methoxyethyl)amino]-5-nitrobenzoic acid

  • MDL number MFCD06655920
  • Cas Number : 854357-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-166
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-13549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13549
Availability details for
Quantity
  • Availability
  • Add to Cart