ENA061488472 Enamine

9-chloro-3,4-dihydro-2H-1,5-benzodioxepine-7-carbonitrile

  • MDL number MFCD06655928
  • Cas Number : 855991-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110...112
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13560

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13560
Availability details for
Quantity
  • Availability
  • Add to Cart