ENA061488478 Enamine

2-(3-bromopropoxy)-1,4-dimethylbenzene

  • MDL number MFCD00078661
  • Cas Number : 3245-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13867

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13867
Availability details for
Quantity
  • Availability
  • Add to Cart