ENA061488486 Enamine

2-[4-(adamantan-1-yl)-1,3-thiazol-2-yl]acetonitrile

  • Cas Number : 497933-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57-58
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13571

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13571
Availability details for
Quantity
  • Availability
  • Add to Cart