ENA061488492 Enamine

2-[2-(thiophen-2-yl)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD06655245
  • Cas Number : 855716-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13575
Availability details for
Quantity
  • Availability
  • Add to Cart