ENA061488496 Enamine

methyl 4-(2,5-dimethoxyphenyl)-2,4-dioxobutanoate

  • MDL number MFCD07324060
  • Cas Number : 70935-15-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85-90
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13885

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13885
Availability details for
Quantity
  • Availability
  • Add to Cart