ENA061488498 Enamine

1-chloro-4-thien-2-ylphthalazine

  • MDL number MFCD06200203
  • Cas Number : 124959-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128...130
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13578

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13578
Availability details for
Quantity
  • Availability
  • Add to Cart