ENA061488499 Enamine

3-{[(furan-2-yl)methyl]amino}-4-hydroxy-1lambda6-thiolane-1,1-dione

  • MDL number MFCD02081102
  • Cas Number : 302806-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141-142
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13888

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13888
Availability details for
Quantity
  • Availability
  • Add to Cart