ENA061488505 Enamine

2-[4-(4-fluorophenyl)-1,3-thiazol-2-yl]acetonitrile

  • MDL number MFCD00107603
  • Cas Number : 342405-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-13581

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13581
Availability details for
Quantity
  • Availability
  • Add to Cart