ENA061488520 Enamine

2-methyl-2-(piperidin-1-yl)propan-1-amine

  • MDL number MFCD01821228
  • Cas Number : 54151-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-13587

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13587
Availability details for
Quantity
  • Availability
  • Add to Cart