ENA061488529 Enamine

2-({3-[(2-chlorobenzyl)oxy]benzyl}amino)ethanol

  • MDL number MFCD03786007
  • Cas Number : 774192-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49...50
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13593

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13593
Availability details for
Quantity
  • Availability
  • Add to Cart