ENA061488533 Enamine

methyl 2-amino-4-(2-chlorophenyl)thiophene-3-carboxylate

  • MDL number MFCD01923581
  • Cas Number : 350997-11-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92-93
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13922

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13922
Availability details for
Quantity
  • Availability
  • Add to Cart