ENA061488535 Enamine

3-(morpholine-4-sulfonyl)propanoic acid

  • MDL number MFCD07323647
  • Cas Number : 848178-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136-137
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13924

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13924
Availability details for
Quantity
  • Availability
  • Add to Cart