ENA061488538 Enamine

2-{[(5-methyl-1,2-oxazol-3-yl)methyl]sulfanyl}pyridine-3-carboxylic acid

  • MDL number MFCD07310146
  • Cas Number : 855991-78-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148-149
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-13600

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13600
Availability details for
Quantity
  • Availability
  • Add to Cart