ENA061488541 Enamine

2-(4-ethoxyphenyl)pyrrolidine

  • MDL number MFCD02663440
  • Cas Number : 383127-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13930

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13930
Availability details for
Quantity
  • Availability
  • Add to Cart