ENA061488551 Enamine

4-(furan-2-yl)-6-oxo-2-sulfanyl-1,6-dihydropyrimidine-5-carbonitrile

  • MDL number MFCD07366328
  • Cas Number : 137446-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-157
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13940

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13940
Availability details for
Quantity
  • Availability
  • Add to Cart