ENA061488559 Enamine

methyl 2,4-dioxo-4-(pyridin-3-yl)butanoate

  • MDL number MFCD06245328
  • Cas Number : 23424-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119-121
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13948

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13948
Availability details for
Quantity
  • Availability
  • Add to Cart