ENA061488581 Enamine

1-[(2,5-difluorophenyl)sulfonyl]piperidine-4-carboxylic acid

  • MDL number MFCD07313708
  • Cas Number : 847759-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13621

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13621
Availability details for
Quantity
  • Availability
  • Add to Cart