ENA061488583 Enamine

1-(1,3-diphenyl-1H-pyrazol-4-yl)methanamine

  • MDL number MFCD02656622
  • Cas Number : 512810-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83-84
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13494

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13494
Availability details for
Quantity
  • Availability
  • Add to Cart