ENA061488587 Enamine

5-{[(furan-2-yl)methyl]amino}-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD07313349
  • Cas Number : 226211-34-3
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124-125
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13626

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13626
Availability details for
Quantity
  • Availability
  • Add to Cart