ENA061488588 Enamine

1-{4-[(5-ethyl-4,5-dihydro-1,3-thiazol-2-yl)amino]phenyl}ethan-1-one

  • MDL number MFCD07314011
  • Cas Number : 854137-76-1
  • Molecular Weight : 248.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-13627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13627
Availability details for
Quantity
  • Availability
  • Add to Cart