ENA061488591 Enamine

3-chloro-5-methoxy-4-propoxybenzonitrile

  • MDL number MFCD02256351
  • Cas Number : 693804-16-9
  • Molecular Weight : 225.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 33-34
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13980

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13980
Availability details for
Quantity
  • Availability
  • Add to Cart