ENA061488592 Enamine

2-(4-ethoxy-3-methoxyphenyl)-2-(1H-indol-3-yl)ethan-1-amine

  • MDL number MFCD06650786
  • Cas Number : 848369-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116-117
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13981

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13981
Availability details for
Quantity
  • Availability
  • Add to Cart