ENA061488604 Enamine

3-[3-sulfanyl-5-(thiophen-2-yl)-4H-1,2,4-triazol-4-yl]propanoic acid

  • MDL number MFCD07312772
  • Cas Number : 854137-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13636

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13636
Availability details for
Quantity
  • Availability
  • Add to Cart