ENA061488608 Enamine

2-{[4-(difluoromethoxy)phenyl]amino}benzoic acid

  • MDL number MFCD01675136
  • Cas Number : 51679-46-0
  • Molecular Weight : 279.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-156
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13997

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13997
Availability details for
Quantity
  • Availability
  • Add to Cart