ENA061488612 Enamine

5-(chloromethyl)-3-(2,4-dimethoxyphenyl)-1,2,4-oxadiazole

  • MDL number MFCD07312768
  • Cas Number : 854137-68-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76-77
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13640

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-13640
Availability details for
Quantity
  • Availability
  • Add to Cart