ENA061957496 Enamine

2-[1-(difluoromethyl)-1H-1,3-benzodiazol-2-yl]-1-(3,4,5-trimethoxyphenyl)ethan-1-one

  • MDL number MFCD07366335
  • Cas Number : 885524-53-8
  • Molecular Weight : 376.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-14066

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14066
Availability details for
Quantity
  • Availability
  • Add to Cart