ENA061957497 Enamine

2-chloro-N-[4-(2-hydroxy-4-methoxyphenyl)-1,3-thiazol-2-yl]acetamide

  • MDL number MFCD07366342
  • Cas Number : 205928-31-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14106

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14106
Availability details for
Quantity
  • Availability
  • Add to Cart