ENA061957500 Enamine

4-ethoxy-3-(morpholine-4-sulfonyl)aniline

  • MDL number MFCD07366345
  • Cas Number : 885524-59-4
  • Molecular Weight : 286.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-14129

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14129
Availability details for
Quantity
  • Availability
  • Add to Cart