ENA061957504 Enamine

2-amino-5-ethoxy-4-methoxybenzoic acid

  • MDL number MFCD07339032
  • Cas Number : 61948-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173-176
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14154

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14154
Availability details for
Quantity
  • Availability
  • Add to Cart