ENA061957510 Enamine

2-(4-oxo-2,3,4,5-tetrahydro-1,5-benzoxazepin-5-yl)acetic acid

  • MDL number MFCD07339053
  • Cas Number : 863668-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188-189
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14162

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14162
Availability details for
Quantity
  • Availability
  • Add to Cart