ENA061957512 Enamine

N-{3-[amino(hydroxyimino)methyl]phenyl}cyclopropanecarboxamide

  • MDL number MFCD07339039
  • Cas Number : 863667-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 229...231
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14168

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14168
Availability details for
Quantity
  • Availability
  • Add to Cart