ENA061957514 Enamine

1-(4-tert-butylphenyl)-2-methylpropan-1-amine

  • MDL number MFCD07339048
  • Cas Number : 863668-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14171

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14171
Availability details for
Quantity
  • Availability
  • Add to Cart