ENA061957524 Enamine

4-(2-methoxyphenyl)-11-methyl-5-sulfanyl-8-thia-4,6-diazatricyclo[7.4.0.0,2,7]trideca-1(9),2(7),5-trien-3-one

  • MDL number MFCD09355774
  • Cas Number : 885524-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14188

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14188
Availability details for
Quantity
  • Availability
  • Add to Cart