ENA061957529 Enamine

2-[(1,1-dioxidotetrahydrothien-3-yl)methyl]-1H-benzimidazole

  • MDL number MFCD07339058
  • Cas Number : 863668-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156...158
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14200

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14200
Availability details for
Quantity
  • Availability
  • Add to Cart