ENA061957532 Enamine

1-(4-methylbenzyl)-3-(4-methylphenyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD07366350
  • Cas Number : 956508-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14204

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14204
Availability details for
Quantity
  • Availability
  • Add to Cart