ENA061957541 Enamine

4-(2,4-dichlorophenyl)-1-(2-methoxyphenyl)-1H-imidazole-2-thiol

  • MDL number MFCD02695895
  • Cas Number : 306956-18-3
  • Molecular Weight : 351.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03157

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03157
Availability details for
Quantity
  • Availability
  • Add to Cart