ENA061957550 Enamine

1H-pyrrole-2-carbohydrazide

  • MDL number MFCD00141885
  • Cas Number : 50269-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 228...230
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14057

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14057
Availability details for
Quantity
  • Availability
  • Add to Cart