ENA061957554 Enamine

N-[4-(aminocarbonothioyl)phenyl]-2-furamide

  • MDL number MFCD07330482
  • Cas Number : 848658-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 258...260
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14061

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14061
Availability details for
Quantity
  • Availability
  • Add to Cart