ENA061957555 Enamine

4-(2-bromoethoxy)benzene-1-sulfonamide

  • MDL number MFCD07329990
  • Cas Number : 125174-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96-97
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14062

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14062
Availability details for
Quantity
  • Availability
  • Add to Cart